2-Chloronicotinonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003361-25G 003361-100G
FLUO003361-25GEA 0 GBP
FLUO003361-25G FLUO003361-100G
2-Chloronicotinonitrile
2-Chloronicotinonitrile
Formula: C₆H₃ClN₂
Boiling Pt: 112 °C (1 mmHg)
Melting Pt: 107…108 °C
MDL Number: MFCD00014628
CAS Number: 6602-54-6
EINECS: 229-550-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR