2-Amino-5-bromopyridine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
5-Bromo-2-pyridylamine, 5-Bromopyridin-2-amine
003417-25G
003417-100G
FLUO003417-25GEA
0
GBP
FLUO003417-25G
FLUO003417-100G
2-Amino-5-bromopyridine
2-Amino-5-bromopyridine
Formula:
C₅H₅BrN₂ Boiling Pt: 244 °C (1013 hPa) Melting Pt: 136…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006323 CAS Number: 1072-97-5 EINECS: 214-019-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...