5-Bromonicotinic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Bromo-5-pyridine carboxylic acid, 5-Bromopyridine-3-carboxylic acid, 5-Bromo-3-pyridinecarboxylic acid
009061-25G
009061-100G
FLUO009061-25GEA
0
GBP
FLUO009061-25G
FLUO009061-100G
5-Bromonicotinic acid
5-Bromonicotinic acid
Formula:
C₆H₄BrNO₂ Boiling Pt: 300 °C (1013 hPa) Melting Pt: 184…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009783 CAS Number: 20826-04-4 EINECS: 244-065-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...