3-Bromo-5-chloropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

033253-5G 033253-1G
FLUO033253-5GEA 0 GBP
FLUO033253-5G FLUO033253-1G
3-Bromo-5-chloropyridine
3-Bromo-5-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
MDL Number: MFCD04114221
CAS Number: 73583-39-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR