3-Chloro-6-methylpyridazine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
637637-1G
637637-5G
SIAL637637-1GEA
45.5
GBP
SIAL637637-1G
SIAL637637-5G
3-Chloro-6-methylpyridazine, Sigma-Aldrich®
3-Chloro-6-methylpyridazine
Formula:
C₅H₅ClN₂ MW: 128.56 g/mol |
MDL Number:
MFCD00052905 CAS Number: 1121-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...