3,6-Dichloro-4-methylpyridazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
CFF121.10
CFF121.50
017102-10G
017102-50G
017102-25G
017102-1G
017102-5G
CHMPCFF121.10EA
0
GBP
CHMPCFF121.10
CHMPCFF121.50
FLUO017102-10G
FLUO017102-50G
FLUO017102-25G
FLUO017102-1G
FLUO017102-5G
3,6-Dichloro-4-methylpyridazine
3,6-Dichloro-4-methylpyridazine
Formula:
C₅H₄Cl₂N₂ Boiling Pt: 130…135 °C (1 mmHg) Melting Pt: 87 °C |
MDL Number:
MFCD00006465 CAS Number: 19064-64-3 EINECS: 242-794-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...