2-Ethyl-3-methylpyrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002731-5G 002731-25G
FLUO002731-5GEA 0 GBP
FLUO002731-5G FLUO002731-25G
2-Ethyl-3-methylpyrazine
2-Ethyl-3-methylpyrazine
Formula: C₇H₁₀N₂
Boiling Pt: 170 °C (1013 hPa)
Density: 0.99 g/cm³ (20 °C)
MDL Number: MFCD00006150
CAS Number: 15707-23-0
EINECS: 239-799-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR