2,6-Dimethylpyrazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

002730-5G 002730-25G
FLUO002730-5GEA 0 GBP
FLUO002730-5G FLUO002730-25G
2,6-Dimethylpyrazine
2,6-Dimethylpyrazine
Formula: C₆H₈N₂
Boiling Pt: 154 °C (1013 hPa)
Melting Pt: 38…42 °C
Density: 0.9647 g/cm³ (50 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00006148
CAS Number: 108-50-9
EINECS: 203-589-4
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR