2,3-Dimethylpyrazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009615-25G
009615-100G
FLUO009615-25GEA
0
GBP
FLUO009615-25G
FLUO009615-100G
2,3-Dimethylpyrazine
2,3-Dimethylpyrazine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 156 Melting Pt: –12 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006144 CAS Number: 5910-89-4 EINECS: 227-630-0 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...