2-Methylpyrazine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
W330906-100G-K
W330906-1KG-K
W330906-5KG-K
SIALW330906-100G-KEA
94.9
GBP
SIALW330906-100G-K
SIALW330906-1KG-K
SIALW330906-5KG-K
2-Methylpyrazine, Sigma-Aldrich®
2-Methylpyrazine
Formula:
C₅H₆N₂ Boiling Pt: 135 °C (1013 hPa) Melting Pt: –30 °C Density: 1.03 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006142 CAS Number: 109-08-0 EINECS: 203-645-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...