2,6-Dichloropyrazine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
002758-25G
002758-100G
FLUO002758-25GEA
0
GBP
FLUO002758-25G
FLUO002758-100G
2,6-Dichloropyrazine
2,6-Dichloropyrazine
Formula:
C₄H₂Cl₂N₂ Boiling Pt: 120…122 °C (40 mmHg) Melting Pt: 57…58 °C Density: 1,713 g/cm³ (20 °C) |
MDL Number:
MFCD00006125 CAS Number: 4774-14-5 EINECS: 225-316-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...