Xanthine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

X4002-1G X4002-25G X4002-5G
SIALX4002-1GEA 80 GBP
SIALX4002-1G SIALX4002-25G SIALX4002-5G
Xanthine, Sigma-Aldrich®
Xanthine
Formula: C₅H₄N₄O₂
MDL Number: MFCD00078453
CAS Number: 69-89-6
EINECS: 200-718-6
Merck Index: 12,10193

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR