Adenine

Supplier: Serva
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

10739.02 10739.03
SERA10739.02EA 40.82 GBP
SERA10739.02 SERA10739.03
Adenine
Adenine
Formula: C₅H₅N₅
Density: 0.99741 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041790
CAS Number: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,00152

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR