tert-Dodecyl mercaptan (mixture of isomers) ≥98.5%, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: tert-Lauryl Mercaptan, tert-Dodecanethiol, Tert-dodecylmercaptan

M-Clarity™ quality level = MQ200

471585-100ML 471585-2L 471585-18L
SIAL471585-100MLEA 80.8 GBP
SIAL471585-100ML SIAL471585-2L SIAL471585-18L
tert-Dodecyl mercaptan (mixture of isomers) ≥98.5%, Sigma-Aldrich®
tert-Dodecyl mercaptan (mixture of isomers)
Formula: C₁₂H₂₆S
MW: 202.40 g/mol
Boiling Pt: 227…248 °C (lit.)
Density: 0.86 g/ml
MDL Number: MFCD00043233
CAS Number: 25103-58-6
UN: 3082
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR