Spironolactone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
S3378-1G
S3378-5G
SIALS3378-1GEA
118
GBP
SIALS3378-1G
SIALS3378-5G
Spironolactone, Sigma-Aldrich®
Spironolactone
Formula:
C₂₄H₃₂O₄S Boiling Pt: 597 °C (1013 hPa) Melting Pt: 196 °C |
MDL Number:
MFCD00082250 CAS Number: 52-01-7 EINECS: 200-133-6 Merck Index: 12,08917 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...