Nickel diiodide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Nickel (II) iodide, Nickel iodide (NiI2)

M-Clarity™ quality level = MQ100

400777-25G 400777-5G
SIAL400777-25GEA 226 GBP
SIAL400777-25G SIAL400777-5G
Nickel diiodide, Sigma-Aldrich®
Nickel diiodide
Formula: I₂Ni
MW: 312.5 g/mol
Melting Pt: 797 °C
Density: 5.83 g/cm³ (20 °C)
MDL Number: MFCD00016264
CAS Number: 13462-90-3
EINECS: 236-666-6
UN: 3260
ADR: 8,II
Merck Index: 12,06594

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR