Cobalt(III) fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

002840-100G 002840-500G
FLUO002840-100GEA 0 GBP
FLUO002840-100G FLUO002840-500G
Cobalt(III) fluoride
Cobalt (III) fluoride
Formula: CoF₃
Melting Pt: 927 °C
Density: 3.88 g/cm³ (20 °C)
MDL Number: MFCD00010942
CAS Number: 10026-18-3
EINECS: 233-062-4
UN: 3260
ADR: 8,II
Merck Index: 12,02491

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR