Cobalt(II) fluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Cobalt difluoride, Cobaltous fluoride
004570-100G
004570-500G
FLUO004570-100GEA
0
GBP
FLUO004570-100G
FLUO004570-500G
Cobalt(II) fluoride
Cobalt (II) fluoride
Formula:
CoF₂ MW: 96.93 g/mol Boiling Pt: 1400 Melting Pt: 1200 Density: 4.46 g/cm³ (20 °C) |
MDL Number:
MFCD00010941 CAS Number: 10026-17-2 EINECS: 233-061-9 UN: 2923 ADR: 8 III Merck Index: 13,02465 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...