Vanadium(V) oxide ≥99.9% (metals basis)
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Vanadium pentoxide, di-Vanadium pentaoxide
44424.14
44424.22
44424.14EA
316
GBP
44424.14
44424.22
Vanadium(V) oxide ≥99.9% (metals basis)
Vanadium (V) oxide
Purity: 99.9%
CAS No.: 1314-62-1
Molecular Formula: V2O5
Formula Weight: 181.88
Physical Form: 3-12mm Pieces
Density: 3.357
Melting Point: 690°
Boiling Point: 1750° dec.
Merck Reference: 14,9921
UN No.: 2862
MDL No.: MFCD00011457
Vacuum deposition
Formula:
O₅V₂ MW: 181.88 g/mol Boiling Pt: 1750 °C (dec.) Melting Pt: 690 °C Density: 3.357 Storage Temperature: Ambient |
MDL Number:
MFCD00011457 CAS Number: 1314-62-1 EINECS: 215-239-8 UN: 2862 ADR: 6.1,III Merck Index: 14,9921 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...