Hafnium(IV) fluoride ≥99.9% (metals basis)
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
45061.14
45061.06
45061.14EA
453
GBP
45061.14
45061.06
Hafnium(IV) fluoride ≥99.9% (metals basis)
Hafnium (IV) fluoride
Purity: 99.9%
CAS No.: 13709-52-9
Molecular Formula: HfF4
Formula Weight: 254.48
Physical Form: -60 Mesh Powder
Sensitivity: Moisture Sensitive
Appearance: White
UN No.: 3290
MDL No.: MFCD00016127
Formula:
F₄Hf Storage Temperature: Ambient |
MDL Number:
MFCD00016127 CAS Number: 13709-52-9 EINECS: 237-258-0 UN: 3290 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...