Zirconium tetrachloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
221880-100G
221880-500G
221880-5G
SIAL221880-100GEA
109
GBP
SIAL221880-100G
SIAL221880-500G
SIAL221880-5G
Zirconium tetrachloride, Sigma-Aldrich®
Zirconium tetrachloride
Formula:
Cl₄Zr Boiling Pt: 3577 °C (1013 hPa) Melting Pt: 437 °C Density: 2.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011306 CAS Number: 10026-11-6 EINECS: 233-058-2 UN: 2503 ADR: 8,III Merck Index: 12,10307 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...