Scandium(III) fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007260-1G 007260-5G
FLUO007260-1GEA 0 GBP
FLUO007260-1G FLUO007260-5G
Scandium(III) fluoride
Scandium (III) fluoride
Formula: F₃Sc
MW: 101.95 g/mol
Boiling Pt: 1527 °C (1013 hPa)
Melting Pt: 1140…1160 °C
Density: 2.52 g/cm³ (25 °C)
MDL Number: MFCD00016325
CAS Number: 13709-47-2
EINECS: 237-255-4
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR