Scandium(III) chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

451266-1G 451266-5G
SIAL451266-1GEA 326 GBP
SIAL451266-1G SIAL451266-5G
Scandium(III) chloride, Sigma-Aldrich®
Scandium (III) chloride
Formula: Cl₃Sc
MW: 151.31 g/mol
Melting Pt: 940 °C
Density: 1.67 g/cm³ (940 °C)
MDL Number: MFCD00011221
CAS Number: 10361-84-9
EINECS: 233-799-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR