Zinc sulphide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Zinc (II) sulphide

M-Clarity™ quality level = MQ200

244627-100G 244627-25G
SIAL244627-100GEA 178 GBP
SIAL244627-100G SIAL244627-25G
Zinc sulphide, Sigma-Aldrich®
Zinc sulphide
Formula: SZn
MW: 97.46 g/mol
Melting Pt: 1064 °C
Density: 4.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011301
CAS Number: 1314-98-3
EINECS: 215-251-3
Merck Index: 12,10294

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR