Zinc oxide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
544906-10G
544906-50G
SIAL544906-10GEA
40.6
GBP
SIAL544906-10G
SIAL544906-50G
Zinc oxide, Sigma-Aldrich®
Zinc oxide
Formula:
ZnO Melting Pt: 1975 °C Density: 5.61 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011300 CAS Number: 1314-13-2 EINECS: 215-222-5 UN: 3077 ADR: 9,III REACH: 01-2119463881-32 Merck Index: 13,10200 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...