Zinc fluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002872-25G
002872-100G
002872-500G
FLUO002872-25GEA
0
GBP
FLUO002872-25G
FLUO002872-100G
FLUO002872-500G
Zinc fluoride
Zinc fluoride
Formula:
F₂Zn MW: 103.39 g/mol Boiling Pt: 1500 Melting Pt: 910…930 °C Density: 4.95 g/cm³ (20 °C) |
MDL Number:
MFCD00011298 CAS Number: 7783-49-5 EINECS: 232-001-9 UN: 3288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...