Mercury(II) fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

002849-25G 002849-100G 002849-250G
FLUO002849-25GEA 0 GBP
FLUO002849-25G FLUO002849-100G FLUO002849-250G
Mercury(II) fluoride
Mercury (II) fluoride
Formula: F₂Hg
Boiling Pt: ∼650 °C (1013 hPa)
Melting Pt: 645 °C
Density: 8.95 g/cm³ (20 °C)
MDL Number: MFCD00016139
CAS Number: 7783-39-3
EINECS: 231-994-6
UN: 2025
ADR: 6.1,II
Merck Index: 12,05930

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR