Copper(II) fluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Cupric fluoride
002841-25G
002841-100G
FLUO002841-25GEA
0
GBP
FLUO002841-25G
FLUO002841-100G
Copper(II) fluoride
Copper (II) fluoride
Formula:
CuF₂ MW: 101.54 g/mol Boiling Pt: 950 Melting Pt: 855 Density: 4.23 g/cm³ (20 °C) |
MDL Number:
MFCD00010977 CAS Number: 7789-19-7 EINECS: 232-147-3 UN: 3288 ADR: 6.1 III Merck Index: 12,02704 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...