Silver carbonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
85150-10G
85150-50G
SIAL85150-10GEA
119
GBP
SIAL85150-10G
SIAL85150-50G
Silver carbonate, Sigma-Aldrich®
Silver carbonate
Formula:
CAg₂O₃ Melting Pt: 210…218 °C Density: 5.65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003403 CAS Number: 534-16-7 EINECS: 208-590-3 Merck Index: 12,08650 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...