Silver chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

449571-1G 449571-5G
SIAL449571-1GEA 115 GBP
SIAL449571-1G SIAL449571-5G
Silver chloride, Sigma-Aldrich®
Silver chloride
Formula: AgCl
MW: 143.32 g/mol
Boiling Pt: 1554
Melting Pt: 455
Density: 5.56 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003399
CAS Number: 7783-90-6
EINECS: 232-033-3
Merck Index: 13,08582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR