Copper(II) bromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Cupric bromide, Cuprous bromide

M-Clarity™ quality level = MQ200

221775-100G 221775-500G 221775-5G
SIAL221775-100GEA 106 GBP
SIAL221775-100G SIAL221775-500G SIAL221775-5G
Copper(II) bromide, Sigma-Aldrich®
Copper (II) bromide
Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900 °C (1013 hPa)
Melting Pt: 498 °C
Density: 4.71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8,III
Merck Index: 13,02656

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR