Gold tribromide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Gold (III) bromide, Gold(III) bromide

M-Clarity™ quality level = MQ100

398470-1G 398470-5G
SIAL398470-1GEA 222 GBP
SIAL398470-1G SIAL398470-5G
Gold tribromide, Sigma-Aldrich®
Gold tribromide
Formula: AuBr₃
Melting Pt: 160 °C
MDL Number: MFCD00014171
CAS Number: 10294-28-7
EINECS: 233-654-2
UN: 3260
ADR: 8,III
Merck Index: 12,04539

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR