Acenaphthene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
|
Synonyms:
1,2-Dihydroacenaphthylene
M-Clarity™ quality level = MQ100
215376-100G
215376-5G
SIAL215376-100GEA
152
GBP
SIAL215376-100G
SIAL215376-5G
Acenaphthene, Sigma-Aldrich®
Acenaphthene
Formula:
C₁₂H₁₀ MW: 154.21 g/mol Boiling Pt: 279 °C (1013 hPa) Melting Pt: 90…93 °C Density: 1.024 g/cm³ (20 °C) Flash Pt: 135 °C (open cup) Storage Temperature: Ambient |
MDL Number:
MFCD00003807 CAS Number: 83-32-9 EINECS: 201-469-6 UN: 3077 ADR: 9,III Merck Index: 13,00029 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...