1-Naphthol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene

M-Clarity™ quality level = MQ200

N1000-100G N1000-10G N1000-500G
SIALN1000-100GEA 118 GBP
SIALN1000-100G SIALN1000-10G SIALN1000-500G
1-Naphthol, Sigma-Aldrich®
1-Naphthol
Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 282
Melting Pt: 94
Density: 1.224 g/cm³ (20 °C)
Flash Pt: 125
Storage Temperature: Ambient
MDL Number: MFCD00003930
CAS Number: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1 III
Merck Index: 13,06409

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR