1-Fluoronaphthalene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

196657-25G 196657-5G
SIAL196657-25GEA 133 GBP
SIAL196657-25G SIAL196657-5G
1-Fluoronaphthalene, Sigma-Aldrich®
1-Fluoronaphthalene
Formula: C₁₀H₇F
MW: 146.16 g/mol
Boiling Pt: 215 °C (1013 hPa)
Melting Pt: –12.5 °C
Density: 1.135 g/cm³ (0 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003873
CAS Number: 321-38-0
EINECS: 206-287-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR