2-Naphthaldehyde, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
β-Naphthaldehyde
M-Clarity™ quality level = MQ200
N206-100G-A
N206-25G-A
N206-5G-A
SIALN206-100G-AEA
345
GBP
SIALN206-100G-A
SIALN206-25G-A
SIALN206-5G-A
2-Naphthaldehyde, Sigma-Aldrich®
2-Naphthaldehyde
Formula:
C₁₁H₈O MW: 156.18 g/mol Boiling Pt: 160 °C (13 mmHg) Melting Pt: 57…60 °C Density: 1.0775 g/cm³ (99 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004094 CAS Number: 66-99-9 EINECS: 200-640-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...