2-Bromonaphthalene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ100

183644-25G 183644-5G
SIAL183644-25GEA 61.9 GBP
SIAL183644-25G SIAL183644-5G
2-Bromonaphthalene, Sigma-Aldrich®
2-Bromonaphthalene
Formula: C₁₀H₇Br
MW: 207.07 g/mol
Boiling Pt: 282
Melting Pt: 52…55 °C
Density: 1.605 g/cm³ (0 °C)
Flash Pt: 112
Storage Temperature: Ambient
MDL Number: MFCD00004051
CAS Number: 580-13-2
EINECS: 209-452-5
Merck Index: 13,01411

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR