Erbium(III) oxide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Dierbium trioxide, Erbium oxide
M-Clarity™ quality level = MQ100
203238-25G
203238-5G
SIAL203238-25GEA
493
GBP
SIAL203238-25G
SIAL203238-5G
Erbium(III) oxide, Sigma-Aldrich®
Erbium (III) oxide
Formula:
Er₂O₃ Melting Pt: 2420 °C Density: 7.36 g/cm³ (20 °C) |
MDL Number:
MFCD00010991 CAS Number: 12061-16-4 EINECS: 235-045-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...