Cerium(IV) oxide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Ceric oxide, Cerium dioxide
22390-100G-F
22390-500G-F
SIAL22390-100G-FEA
83.3
GBP
SIAL22390-100G-F
SIAL22390-500G-F
Cerium(IV) oxide, Sigma-Aldrich®
Cerium (IV) oxide
Formula:
CeO₂ MW: 172.11 g/mol Boiling Pt: 3468 Melting Pt: 2000 Density: 7.13 g/cm³ (25 °C) |
MDL Number:
MFCD00010933 CAS Number: 1306-38-3 EINECS: 215-150-4 Merck Index: 12,02035 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...