Terbium(III) fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Terbium(III) fluoride, Terbium trifluoride

007305-5G 007305-25G
FLUO007305-5GEA 0 GBP
FLUO007305-5G FLUO007305-25G
Terbium(III) fluoride
Terbium (III) fluoride
Formula: TbF₃
Boiling Pt: 2277 °C (1013 hPa)
Melting Pt: 1172 °C
Density: 7.23 g/cm³ (20 °C)
MDL Number: MFCD00049572
CAS Number: 13708-63-9
EINECS: 237-247-0
UN: 3288
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR