Neodymium trifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
006868-10G
006868-100G
006868-500G
FLUO006868-10GEA
0
GBP
FLUO006868-10G
FLUO006868-100G
FLUO006868-500G
Neodymium trifluoride
Neodymium trifluoride
Formula:
F₃Nd Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 1410 °C Density: 6.5 g/cm³ (20 °C) |
MDL Number:
MFCD00011135 CAS Number: 13709-42-7 EINECS: 237-253-3 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...