Europium(III) fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006156-1G 006156-5G
FLUO006156-1GEA 0 GBP
FLUO006156-1G FLUO006156-5G
Europium(III) fluoride
Europium (III) fluoride
Formula: EuF₃
Boiling Pt: 2280 °C (1013 hPa)
Melting Pt: 1249.85…1265.85 °C
MDL Number: MFCD00010998
CAS Number: 13765-25-8
EINECS: 237-368-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR