Samarium trichloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Samarium (III) chloride

M-Clarity™ quality level = MQ100

400610-10G 400610-2G
SIAL400610-10GEA 255 GBP
SIAL400610-10G SIAL400610-2G
Samarium trichloride, Sigma-Aldrich®
Samarium trichloride
Formula: Cl₃Sm
MW: 256.72 g/mol
Melting Pt: 668 °C
Density: 4.27 g/cm³ (0 °C)
MDL Number: MFCD00011235
CAS Number: 10361-82-7
EINECS: 233-797-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR