1-Methylindole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-Methylindole, 1-Methyl-1H-indole

193984-25G 193984-5G
SIAL193984-25GEA 60.3 GBP
SIAL193984-25G SIAL193984-5G
1-Methylindole, Sigma-Aldrich®
1-Methylindole
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 239 °C (1013 hPa)
Melting Pt: 94…95 °C
Density: 1.048 g/cm³ (20 °C)
MDL Number: MFCD00005800
CAS Number: 603-76-9
EINECS: 210-057-5
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR