7-Nitroindole ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

A19384.03 A19384.06 A19384.14
A19384.03EA 59.8 GBP
A19384.03 A19384.06 A19384.14
7-Nitroindole ≥97%
7-Nitroindole

Purity: 97% CAS No.: 6960-42-5 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Odour: Odourless Melting Point: 94-96° Beilstein Reference: 130874 MDL No.: MFCD00005683

Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 94…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00005683
CAS Number: 6960-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR