6-Nitroindole ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

A12927.03 A12927.14
A12927.03EA 27.1 GBP
A12927.03 A12927.14
6-Nitroindole ≥97%
6-Nitroindole

Purity: 97% CAS No.: 4769-96-4 Molecular Formula: C8H6N2O2 Formula Weight: 162.15 Melting Point: 141-145° Beilstein Reference: 135977

Formula: C₈H₆N₂O₂
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00051497
CAS Number: 4769-96-4
EINECS: 225-311-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR