6-Fluoroindole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Fluoro-1H-indole

005410-1G 005410-5G
FLUO005410-1GEA 0 GBP
FLUO005410-1G FLUO005410-5G
6-Fluoroindole
6-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00056933
CAS Number: 399-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR