7-Bromoindole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 7-Bromo-1H-indole

MQ100

473723-1G 473723-5G
SIAL473723-1GEA 152 GBP
SIAL473723-1G SIAL473723-5G
7-Bromoindole, Sigma-Aldrich®
7-Bromoindole
Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD00799492
CAS Number: 51417-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR