6-Bromoindole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromo-1H-indole

013449-1G 013449-5G
FLUO013449-1GEA 0 GBP
FLUO013449-1G FLUO013449-5G
6-Bromoindole
6-Bromoindole
Formula: C₈H₆BrN
MW: 196.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238550
CAS Number: 52415-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR