5-Chloro-1-indanone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001148-5G
001148-25G
FLUO001148-5GEA
0
GBP
FLUO001148-5G
FLUO001148-25G
5-Chloro-1-indanone
5-Chloro-1-indanone
Formula:
C₉H₇ClO MW: 166.61 g/mol Boiling Pt: 150 °C (0.2 mmHg) Melting Pt: 100 |
MDL Number:
MFCD00041456 CAS Number: 42348-86-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...